missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM7 Antibody (MCM7/1468), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59611JF549
This item is not returnable.
View return policy
Description
MCM7 Monoclonal antibody specifically detects MCM7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Specifications
| MCM7 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47 | |
| Recombinant human MCM7 protein fragment (aa195-319) (Uniprot: P33993) | |
| 0.1 mL | |
| Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4176 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF | |
| MCM7/1468 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction