Learn More
MCM7 Antibody (MCM7/2756R), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-08404AF750
Code produit. 18553830
  • € 621.00 / 100µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
MCM7 Monoclonal antibody specifically detects MCM7 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
MCM7
Monoclonal
Alexa Fluor 750
CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47
Recombinant fragment of human MCM7 (aa195-319) (Uniprot: P33993)
100 μg
Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers
4176
Store at 4°C in the dark.
IgG
Western Blot, Immunohistochemistry (Paraffin)
MCM7/2756R