missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM7 Antibody (MCM7/2832R), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08405F
This item is not returnable.
View return policy
Description
MCM7 Monoclonal antibody specifically detects MCM7 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| MCM7 | |
| Monoclonal | |
| FITC | |
| CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47 | |
| Recombinant human MCM7 protein fragment (aa195-319) (Uniprot: P33993) | |
| 100 μg | |
| Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4176 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| MCM7/2832R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction