missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCM8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599829
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MCM8.
The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein contains the central domain that is conserved among the MCM proteins. This protein has been shown to co-immunoprecipitate with MCM4, 6 and 7, which suggests that it may interact with other MCM proteins and play a role in DNA replication. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
| MCM8 | |
| Polyclonal | |
| Unconjugated | |
| MCM8 | |
| 5730432L01Rik; C20orf154; dJ967N21.5; DNA helicase MCM8; DNA replication licensing factor MCM8; MCM8; MCM8 minichromosome maintenance deficient 8; MGC119522; MGC119523; MGC12866; MGC4816; minichromosome maintenance 8; minichromosome maintenance 8 homologous recombination repair factor; minichromosome maintenance complex component 8; minichromosome maintenance deficient 8; minichromosome maintenance deficient 8 (S. cerevisiae); POF10; REC; REC homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 296178, 84515 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| D3ZVK1, Q9UJA3 | |
| MCM8 | |
| A synthesized peptide derived from human MCM8(Accession Q9UJA3), corresponding to amino acid residues K734-N784. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction