Learn More
Invitrogen™ MCMBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110378
Description
Immunogen sequence: MPCGEDWLSH PLGIVQGFFA QNGVNPDWEK KVIEYFKEKL KENNAPKWVP SLNEVPLHYL KPNSFVKFRC MIQDMFDPEF YMGVYETVNQ NTKAHVLHFG KYRDVAECGP QQELDLNSPR NTTLERQTFY CVPVPGESTW VKEAYVNANQ ARVSPSTSYT PSRHKRSYED DDDMDLQPNK QKDQHAGARQ AGSVGGLQWC GEPKRLETEA STGQQLNSLN LSSPFDLNFP LPGEKGPACL.
MEK1 (Mitogen activated protein kinase kinase 1) catalyzes the concomitant phosphorylation of a threonine and a tyrosine residue in a Thr-Glu-Tyr sequence located in MAP kinases. MEK1 activates ERK1 and ERK2 MAP kinases. Mitogen activated protein kinase kinase 2 (MEK2 or MAPKK2) is a member of a family of tyrosine/threonine protein kinases that activate the ERK1 and 2 and MAPK enzymes by phosphorylating both residues within the threonine/glutamate/tyrosine (TEY) motif in the activation loop. MEK1 and 2 are also activated by dual phosphorylation, which occurs on serine 218 and 222, in the activation loop of the MEK. Threonine 292 of MEK1 is phosphorylated by ERK 2, which serves as a negative feedback loop by suppressing activation of MEK1.
Specifications
| MCMBP | |
| Polyclonal | |
| Unconjugated | |
| MCMBP | |
| 1110007A13Rik; AU017382; BC025641; C10orf119; cj119; MCM (minichromosome maintenance deficient) binding protein; MCM-binding protein; Mcmbp; MCM-BP; minichromosome maintenance complex binding protein; Mini-chromosome maintenance complex-binding protein; minichromosome maintenance complex-binding protein; RGD1306730 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 210711, 79892 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.96 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8R3C0, Q9BTE3 | |
| MCMBP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human MCMBP (NP_079110.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.