missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCP-1 Monoclonal Antibody (KT221)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549469
This item is not returnable.
View return policy
Description
This product was used in both Indirect and Sandwich ELISA. This product does not cross-react with human IL1B, IL2, IL4, IL5, IL6, IL7, CXCL8, IL9, IL10, IL11, IL12, IL13, CSF2, IFNA, IFNG, KITLG and TNF. This product can be paired with MA5-49467.
This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
Specifications
| MCP-1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P13500 | |
| Ccl2 | |
| Recombinant Human CCL2 (aa 31-90). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| KT221 | |
| Unconjugated | |
| Ccl2 | |
| Acidic seminal fluid protein; AI323594; C-C motif chemokine 2; C-C motif chemokine ligand 2; Ccl2; chemokine (C-C motif) ligand 2; GDCF-2; HC11; H-MCP-1; HSMCR30; immediate-early serum-responsive JE protein; immediate-early serum-responsive protein JE; Je; MCAF; Mcp1; MCP-1; MCP1A; MCP-1A; MGC9434; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; monocyte chemotactic protein 1A; Monocyte secretory protein JE; monocyte-chemoattractant protein-1 precursor; platelet-derived growth factor-inducible protein JE; RP23-350G1.3; SCYA2; Sigje; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; small inducible gene JE; small-inducible cytokine A2; SMC-CF | |
| Mouse | |
| Protein A | |
| RUO | |
| 6347 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction