missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCPH1 Monoclonal Antibody (OTI2A3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526706
This item is not returnable.
View return policy
Description
MCPH1 Monoclonal Antibody for Western Blot, IHC (P)
This protein is implicated in chromosome condensation and DNA damage induced cellular responses, and may play a role in neurogenesis and regulation of the size of the cerebral cortex.
Specifications
| MCPH1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NEM0 | |
| MCPH1 | |
| Human recombinant protein fragment corresponding to amino acids 1-264 of MCPH1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A3 | |
| Unconjugated | |
| MCPH1 | |
| 5430437K10Rik; BRCT-repeat inhibitor of TERT expression 1; BRIT 1; BRIT1; D030046N04Rik; FLJ12847; Mcph1; MCT; Microcephalin; microcephalin 1; microcephaly, primary autosomal recessive 1; RGD1560642; truncated microcephalin | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79648 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto