missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCRS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11362-1-AP-20UL
This item is not returnable.
View return policy
Description
Modulates the transcription repressor activity of DAXX by recruiting it to the nucleolus. As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues. Putative regulatory component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair. May also be an inhibitor of TERT telomerase activity.Specifications
| MCRS1 | |
| Polyclonal | |
| Unconjugated | |
| MCRS1 | |
| 58 kDa microspherule protein, ICP22BP, INO80 complex subunit J, INO80Q, MCRS1, MCRS2, microspherule protein 1, MSP58, P78 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10445, 300222, 51812 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96EZ8, Q99L90 | |
| Mcrs1 | |
| MCRS1 Fusion Protein Ag1904 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction