missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MCT3/SLC16A8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 213.00 - € 508.00
Specifications
| Antigen | MCT3/SLC16A8 |
|---|---|
| Dilution | Western Blot : 1-5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066866
|
Novus Biologicals™
NBP3-30934-100ug |
100 μg |
€ 508.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066873
|
Novus Biologicals™
NBP3-30934-25ug |
25 μg |
€ 213.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCT3/SLC16A8 Polyclonal antibody specifically detects MCT3/SLC16A8 in Human samples. It is validated for Western BlotSpecifications
| MCT3/SLC16A8 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| MCT3MCT 3, monocarboxylate transporter 3, REMP, solute carrier 16 (monocarboxylic acid transporters), member 8, Solute carrier family 16 member 8, solute carrier family 16, member 8 (monocarboxylic acid transporter 3) | |
| Synthetic peptide derived from MCT3/SLC16A8 protein. | |
| Ammonium sulfate precipitation |
| Western Blot : 1-5 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline | |
| 23539 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title