missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCT4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | MCT4 |
|---|---|
| Concentration | 0.38 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16882085
|
Proteintech
22787-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16872085
|
Proteintech
22787-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Minus end-directed microtubule-dependent motor required for bipolar spindle formation. May contribute to movement of early endocytic vesicles. Regulates cilium formation and structure.Specifications
| MCT4 | |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| O15427, P57787 | |
| 80879, 9123 | |
| MCT4 Fusion Protein Ag18788 | |
| Primary | |
| -20°C | |
| SLC16A3 |
| 0.38 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| MCT 4, MCT4, Monocarboxylate transporter 3, Monocarboxylate transporter 4, SLC16A3 | |
| SLC16A3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title