Learn More
MCTS1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-128AF647
This item is not returnable.
View return policy
Description
MCTS1 Polyclonal antibody specifically detects MCTS1 in Human, Mouse samples. It is validated for Western Blot