missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCTS1 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-128IR
This item is not returnable.
View return policy
Description
MCTS1 Polyclonal antibody specifically detects MCTS1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| MCTS1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 755 | |
| malignant T cell amplified sequence 1, Malignant T-Cell-Amplified Sequence 1, MCT-1, Multiple Copies T-Cell Malignancies | |
| Partial residue sequence of the 20 kDa MCT1 protein | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 28985 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction