missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MD-1/LY86 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76709MFV500
This item is not returnable.
View return policy
Description
MD-1/LY86 Polyclonal antibody specifically detects MD-1/LY86 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MD-1/LY86 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Ly-86, lymphocyte antigen 86, MD1, MMD-1, Protein MD-1, RP105-associated | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the center of human MD-1. The immunogen is located within the last 50 amino acids of MD-1. Amino Acid Squence: CGRRKGEQIYYAGPV | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Immunology | |
| 9450 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction