missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MD-1 Monoclonal Antibody (153014)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA562012
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
MD-1, also known as LY86, is a glycoprotein involved in innate immunity. It is a component of the RP105/MD-1 complex, which is expressed primarily in immune cells like B-cells. MD-1 is structurally similar to MD-2, sharing only 16% sequence identity, yet both are implicated in lipopolysaccharide recognition. The protein contains the ML (MD-2-related lipid-recognition) domain, which is conserved across various species and mediates diverse biological functions by interacting with specific lipids. Structurally, MD-1 is characterized by a beta-rich fold consisting of multiple strands, which is integral to its biological activities. Though MD-1 mainly associates with RP105, its role in lipid recognition suggests functional similarities with proteins such as GM2A and NPC2, implying participation in broader immune signaling pathways.
Specifications
| MD-1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O95711 | |
| LY86 | |
| Mouse myeloma cell line NS0-derived recombinant human MD-1, Gly21-Ser162. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 153014 | |
| Unconjugated | |
| LY86 | |
| dJ80N2.1; Ly86; Ly-86; Lymphocyte antigen 86; MD1; MD-1; MD-1, RP105-associated; MMD-1; Protein MD-1; RP1-80N2.1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 9450 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction