missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MD-1 Polyclonal Antibody

Description
Reconstitute at 0.2 mg/mL in sterile PBS.
MD-1, also known as LY86, is a glycoprotein involved in innate immunity. It is a component of the RP105/MD-1 complex, which is expressed primarily in immune cells like B-cells. MD-1 is structurally similar to MD-2, sharing only 16% sequence identity, yet both are implicated in lipopolysaccharide recognition. The protein contains the ML (MD-2-related lipid-recognition) domain, which is conserved across various species and mediates diverse biological functions by interacting with specific lipids. Structurally, MD-1 is characterized by a beta-rich fold consisting of multiple strands, which is integral to its biological activities. Though MD-1 mainly associates with RP105, its role in lipid recognition suggests functional similarities with proteins such as GM2A and NPC2, implying participation in broader immune signaling pathways.
Specifications
Specifications
| Antigen | MD-1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with trehalose and no preservative |
| Gene | LY86 |
| Gene Accession No. | O88188 |
| Gene Alias | dJ80N2.1; Ly86; Ly-86; Lymphocyte antigen 86; MD1; MD-1; MD-1, RP105-associated; MMD-1; Protein MD-1; RP1-80N2.1 |
| Gene Symbols | LY86 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?