Learn More
MD-2 Antibody (1A2E3), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-75513PCP
Product Code. 18506182
  • € 599.00 / 0.10mL
Description
MD-2 Monoclonal antibody specifically detects MD-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MD-2
Monoclonal
PerCP
PBS
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
1A2E3
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
ESOP1, ESOP-1, Ly-96, lymphocyte antigen 96, MD2, MD-2, myeloid differentiation protein-2, Protein MD-2
Antibody was raised against a recombinant protein corresponding to amino acids 2 to 160 of human MD-2.
0.1 mL
Signal Transduction
23643
Store at 4C in the dark.