missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MD-2 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77201AF405
This item is not returnable.
View return policy
Description
MD-2 Polyclonal antibody specifically detects MD-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MD-2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| ESOP1, ESOP-1, Ly-96, lymphocyte antigen 96, MD2, MD-2, myeloid differentiation protein-2, Protein MD-2 | |
| Antibody was raised against a peptide corresponding to 13 amino acids near the center of human MD-2. The immunogen is located within the last 50 amino acids of MD-2. Amino Acid Sequence: CSFKGIKFSKGKYK | |
| 0.1 mL | |
| Signal Transduction | |
| 23643 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction