missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDA5 Antibody (3F6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00064135-M02
This item is not returnable.
View return policy
Description
MDA5 Monoclonal antibody specifically detects MDA5 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Specifications
| MDA5 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| CADM-140 autoantigen, Clinically amyopathic dermatomyositis autoantigen 140 kDa, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, EC 3.6.4.13, Helicard, Helicase with 2 CARD domains, Hlcd, IDDM19, interferon induced with helicase C domain 1, interferon-induced helicase C domain-containing protein 1, Interferon-induced with helicase C domain protein 1, MDA-5MGC133047, MDA5RNA helicase-DEAD box protein 116, melanoma differentiation associated protein-5, Melanoma differentiation-associated protein 5, Murabutide down-regulated protein, RH116 | |
| IFIH1 (NP_071451.2, 928 a.a. ∽ 1023 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. HVNMTPEFKELYIVRENKALQKKCADYQINGEIICKCGQAWGTMMVHKGLDLPCLKIRNFVVVFKNNSTKKQYKKWVELPITFPNLDYSECCLFSD | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 64135 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA | |
| 3F6 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA | |
| NP_071451 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction