missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDA5 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76761AF647
This item is not returnable.
View return policy
Description
MDA5 Polyclonal antibody specifically detects MDA5 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| MDA5 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 647 | |
| CADM-140 autoantigen, Clinically amyopathic dermatomyositis autoantigen 140 kDa, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, EC 3.6.4.13, Helicard, Helicase with 2 CARD domains, Hlcd, IDDM19, interferon induced with helicase C domain 1, interferon-induced helicase C domain-containing protein 1, Interferon-induced with helicase C domain protein 1, MDA-5MGC133047, MDA5RNA helicase-DEAD box protein 116, melanoma differentiation associated protein-5, Melanoma differentiation-associated protein 5, Murabutide down-regulated protein, RH116 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the center of human MDA5. The immunogen is located within amino acids 330 - 380 of MDA5. Amino Acid Squence: AKDHLDKKKKASEPGK | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 64135 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction