missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MDC Monoclonal Antibody (57226)

Mouse Monoclonal Antibody
€ 457.00 - € 688.00
Specifications
| Antigen | MDC |
|---|---|
| Clone | 57226 |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Neutralization, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15573442
|
Invitrogen™
MA523754 |
500 μg |
€ 688.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15583522
![]() |
Invitrogen™
MA523835 |
100 μg |
€ 457.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In ELISAs, this antibody does notcross-react with recombinant mouse (rm) CCL17, rhCCL17, or rhCCL21. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
CCL22 may play a role in the trafficking of activated/effector T-lymphocytes to inflammatory sites and other aspects of activated T-lymphocyte physiology. Also, it is a chemotactic for monocytes. dendritic cells and natural killer cells. CCL22 is a mild chemoattractant for primary activated T-lymphocytes and a potent chemoattractant for chronically activated T-lymphocytes but has no chemattractant activity for neutrophils, eosinophils, and resting T-lymphocytes. Binds to CCR4.Specifications
| MDC | |
| 0.5 mg/mL | |
| ELISA, Neutralization, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 5% trehalose and No Preservative | |
| O00626 | |
| 6367 | |
| E. coli-derived recombinant human CCL22/MDC Gly25-Gln93. | |
| Antibody |
| 57226 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Monoclonal | |
| Lyophilized | |
| IgG2b | |
| Human | |
| CCL22 | |
| A-152E5.1; Abcd1; ABCD-1; Activated B and dendritic cell-derived; AW060611; CC chemokine ABCD-1; CC chemokine STCP-1; C-C motif chemokine 22; C-C motif chemokine ligand 22; Ccl22; chemokine (C-C motif) ligand 22; DC/B-CK; DCBCK; dendritic cell and B cell derived chemokine; macrophage-derived chemokine; MDC; MDC(1-69); MDC(3-69); MDC(5-69); MDC(7-69); MGC108943; MGC34554; RP24-69A20.1; Scya22; small inducible cytokine A22; SMALL INDUCIBLE CYTOKINE A22 PRECURSOR (CC CHEMOKINE ABCD-1) (ACTIVATED B AND DENDRITIC CELL-DERIVED); small inducible cytokine subfamily A (Cys-Cys) member 22; small inducible cytokine subfamily A (Cys-Cys), member 22; small inducible cytokine subfamily A, member 22; small inducible cytokine subfamily A22; small-inducible cytokine A22; STCP-1; stimulated T cell chemotactic protein 1; stimulated T-cell chemotactic protein 1 | |
| CCL22 | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title