missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDC1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-395C
This item is not returnable.
View return policy
Description
MDC1 Polyclonal specifically detects MDC1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| MDC1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| DKFZp781A0122, Em:AB023051.5, KIAA0170MGC166888, mediator of DNA damage checkpoint 1, mediator of DNA damage checkpoint protein 1, mediator of DNA-damage checkpoint 1, NFBD1homologue to Drosophila photoreceptor protein calphotin, Nuclear factor with BRCT domains 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MDC1 | |
| A synthetic peptide, which represented a portion of human Mediator of DNA Damage Checkpoint Protein 1 encoded within exon 5 (NP_055456.1). | |
| 100 μL | |
| Autophagy, DNA Repair | |
| 9656 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction