missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 17 publications
Brand: Novus Biologicals NB100-395
This item is not returnable.
View return policy
Description
MDC1 Polyclonal specifically detects MDC1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| MDC1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DKFZp781A0122, Em:AB023051.5, KIAA0170MGC166888, mediator of DNA damage checkpoint 1, mediator of DNA damage checkpoint protein 1, mediator of DNA-damage checkpoint 1, NFBD1homologue to Drosophila photoreceptor protein calphotin, Nuclear factor with BRCT domains 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Based on 100% sequence identity, this antibody is predicted to react with Rhesus Monkey, Gorilla and Chimpanzee. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:250, Immunoprecipitation 2 - 5 ug/mg lysate | |
| Q14676 | |
| MDC1 | |
| A synthetic peptide, which represented a portion of human Mediator of DNA Damage Checkpoint Protein 1 encoded within exon 5 (NP_055456.1). | |
| 100 μL | |
| Autophagy, DNA Repair | |
| 9656 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction