missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-397
This item is not returnable.
View return policy
Description
MDC1 Polyclonal specifically detects MDC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| MDC1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DKFZp781A0122, Em:AB023051.5, KIAA0170MGC166888, mediator of DNA damage checkpoint 1, mediator of DNA damage checkpoint protein 1, mediator of DNA-damage checkpoint 1, NFBD1homologue to Drosophila photoreceptor protein calphotin, Nuclear factor with BRCT domains 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB 100-397 recognizes human MDC1. Based on 100% sequence identity, this antibody is predicted to react with Chimpanzee. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:5000-1:25000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:250 - 1:1000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000 | |
| Q14676 | |
| MDC1 | |
| The immunogen this antibody was made to, maps to a region between residues 1375 and 1425 of human Mediator of DNA Damage Checkpoint 1 using the numbering given in entry NP_055456.1 (GeneID 9656). | |
| 0.1 mL | |
| Autophagy, DNA Repair | |
| 9656 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction