missing translation for 'onlineSavingsMsg'
Learn More

MDC1 Antibody (P2B11), Novus Biologicals™

Mouse Monoclonal Antibody

€ 179.00

Specifications

Antigen MDC1
Clone P2B11
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Applications Western Blot, Immunocytochemistry, Immunofluorescence
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18723384
View Documents
Novus Biologicals
NBP2-12890-0.025mg
0.025 mg
€ 179.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MDC1 Monoclonal specifically detects MDC1 in Human, Mouse, Bovine, Non-human Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

MDC1
1 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Mouse
9656
GST-tagged recombinant protein corresponding to mouse MDC1 at and around the N-terminus
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
P2B11
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Monoclonal
Purified
RUO
DKFZp781A0122, Em:AB023051.5, KIAA0170MGC166888, mediator of DNA damage checkpoint 1, mediator of DNA damage checkpoint protein 1, mediator of DNA-damage checkpoint 1, NFBD1homologue to Drosophila photoreceptor protein calphotin, Nuclear factor with BRCT domains 1
MDC1
IgG1
Protein G purified
Detects approx 184kDa. This antibody recognizes MDC1 at and around the N-terminus.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.