missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MDGA1 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547691
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human MDGA2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
| MDGA1 | |
| Polyclonal | |
| Unconjugated | |
| MDGA1 | |
| FLJ45018; glycosyl-phosphatidyl-inositol-MAM; glycosylphosphatidylinositol-MAM; GPI and MAM protein; GPIM; MAM domain containing glycosylphosphatidylinositol anchor 1; MAM domain-containing glycosylphosphatidylinositol anchor protein 1; MAM domain-containing protein 3; MAMDC3; MDGA1 | |
| Sheep | |
| Antigen Affinity Chromatography | |
| RUO | |
| 266727 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8NFP4 | |
| MDGA1 | |
| Mouse myeloma cell line NS0-derived recombinant human MDGA1 Gln19-Gly931. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction