missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDGA2 Antibody (803732R), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5184RH
This item is not returnable.
View return policy
Description
MDGA2 Monoclonal antibody specifically detects MDGA2 in Human, Rat samples. It is validated for Flow Cytometry, CyTOF
Specifications
| MDGA2 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 803732R | |
| Flow Cytometry, CyTOF-ready | |
| MAM domain containing glycosylphosphatidylinositol anchor 2 | |
| Mouse myeloma cell line NS0-derived recombinant human MDGA2, Gln21-Asp934, Accession # Q7Z553 | |
| RUO | |
| 161357 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction