missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
€ 422.00 - € 423.00
Specifications
Antigen | MDH1 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16530792
|
Invitrogen
PA597964 |
100 μg |
€ 423.00
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16540792
|
Invitrogen
PA597965 |
100 μL |
€ 422.00
100µL |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.Specifications
MDH1 | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
B230377B03Rik, Cytosolic malate dehydrogenase, D17921, Diiodophenylpyruvate reductase, epididymis secretory protein Li 32, HEL-S-32, malate dehydrogenase (EC 1.1.1.37), malate dehydrogenase 1, malate dehydrogenase 1, NAD (soluble), malate dehydrogenase, cytoplasmic, malate dehydrogenase, cytoplasmic, malate dehydrogenase, peroxisomal, malate dehydrogenase, soluble, Malate dehydrogenase-like enzyme, Mdh, Mdh1, MDHA, Mdhl, MDH-s, MDL1, MGC:1375, Mor2, Mor-2, soluble malate dehydrogenase | |
MDH1 | |
IgG | |
-20°C or -80°C if preferred | |
MDH1 |
Polyclonal | |
Liquid | |
RUO | |
P14152, P40925 | |
17449, 4190 | |
Recombinant Human Malate dehydrogenase, cytoplasmic protein (1-334AA) | |
Primary | |
Antibody |