missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MDH1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143035
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize all reported isoforms (NP_001186040.1; NP_005908.1; NP_001186041.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.
Specifications
| MDH1 | |
| Polyclonal | |
| Unconjugated | |
| MDH1 | |
| B230377B03Rik; Cytosolic malate dehydrogenase; D17921; Diiodophenylpyruvate reductase; epididymis secretory protein Li 32; HEL-S-32; malate dehydrogenase (EC 1.1.1.37); malate dehydrogenase 1; malate dehydrogenase 1, NAD (soluble); malate dehydrogenase, cytoplasmic; malate dehydrogenase, cytoplasmic; malate dehydrogenase, peroxisomal; malate dehydrogenase, soluble; Malate dehydrogenase-like enzyme; Mdh; Mdh1; MDHA; Mdhl; MDH-s; MDL1; MGC:1375; Mor2; Mor-2; soluble malate dehydrogenase | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 17449, 24551, 396894, 4190 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O88989, P11708, P14152, P40925 | |
| MDH1 | |
| Peptide with sequence C-PDVNHAKVKLQGK (aa 193-205). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction