missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MDH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550446
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total MDH1 protein.
MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.
Specifications
| MDH1 | |
| Polyclonal | |
| Unconjugated | |
| MDH1 | |
| B230377B03Rik; Cytosolic malate dehydrogenase; D17921; Diiodophenylpyruvate reductase; epididymis secretory protein Li 32; HEL-S-32; malate dehydrogenase (EC 1.1.1.37); malate dehydrogenase 1; malate dehydrogenase 1, NAD (soluble); malate dehydrogenase, cytoplasmic; malate dehydrogenase, cytoplasmic; malate dehydrogenase, peroxisomal; malate dehydrogenase, soluble; Malate dehydrogenase-like enzyme; Mdh; Mdh1; MDHA; Mdhl; MDH-s; MDL1; MGC:1375; Mor2; Mor-2; soluble malate dehydrogenase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17449, 4190 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 2.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P14152, P40925 | |
| MDH1 | |
| Fusion protein corresponding to a region derived from internal residues of human malate dehydrogenase 1, NAD. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction