missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MDH2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143303
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
Malate dehydrogenase catalyzes the reversible oxidation of malate to oxaloacetate, utilizing the NAD/NADH cofactor system in the citric acid cycle. The protein encoded by this gene is localized to the mitochondria and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria. Several transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2013].
Specifications
| MDH2 | |
| Polyclonal | |
| Unconjugated | |
| MDH2 | |
| malate dehydrogenase 2; malate dehydrogenase 2, NAD (mitochondrial); malate dehydrogenase precursor (EC 1.1.1.37); malate dehydrogenase, mitochondrial; MDH; Mdh2; Mdh-2; MGC:3559; mitochondrial malate dehydrogenase 2, NAD; MMDH; M-MDH; Mor1; Mor-1; testicular tissue protein Li 120 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 17448, 397039, 4191, 81829 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P00346, P04636, P08249, P40926 | |
| MDH2 | |
| Peptide with sequence C-TRDDLFNTNAT. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction