Learn More
MDK Monoclonal Antibody (OTI6C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807679
This item is not returnable.
View return policy
Description
Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis.Specifications
| MDK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P21741 | |
| MDK | |
| Human recombit protein fragment corresponding to amino acids 21-143 of human MDK produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6C8 | |
| Unconjugated | |
| MDK | |
| ARAP, MK, NEGF2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4192 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |