Learn More
MDK Monoclonal Antibody (OTI6C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA807679
Encompass_Preferred
 View more versions of this product
Product Code. 17568353
  • € 540.87 / 100µL
Description
Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis.
Specifications
Specifications
MDK
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P21741
MDK
Human recombit protein fragment corresponding to amino acids 21-143 of human MDK produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI6C8
Unconjugated
MDK
ARAP, MK, NEGF2
Mouse
Affinity Chromatography
RUO
4192
-20° C, Avoid Freeze/Thaw Cycles