missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDL-1/CLEC5A Antibody (226402), DyLight 680, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1639Y
This item is not returnable.
View return policy
Description
MDL-1/CLEC5A Monoclonal antibody specifically detects MDL-1/CLEC5A in Mouse samples. It is validated for Flow Cytometry
Specifications
| MDL-1/CLEC5A | |
| Monoclonal | |
| DyLight 680 | |
| CLECSF5carbohydrate-recognition domain) lectin, superfamilymember 5, C-type lectin domain family 5, member A, C-type lectin superfamily member 5, MDL-1MGC138304, Myeloid DAP12-associating lectin 1, myeloid DAP12-associating lectin-1 | |
| BaF3 mouse pro-B cell line transfected with mouse MDL-1/CLEC5A | |
| 0.5 mL | |
| Immunology | |
| 23601 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| 226402 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction