missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 292.00 - € 590.10
Specifications
| Antigen | MDM1 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18459750
|
Novus Biologicals
NBP1-86242-25ul |
25 μL |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18280897
|
Novus Biologicals
NBP1-86242 |
0.1 mL |
€ 624.00 € 590.10 / 0.10mL Save € 33.90 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MDM1 Polyclonal specifically detects MDM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MDM1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ95264, Mdm1 nuclear protein homolog (mouse), Mdm4, transformed 3T3 cell double minute 1, p53 binding protein, Mdm4, transformed 3T3 cell double minute 1, p53 binding protein (mouse), nuclear protein double minute 1, nuclear protein MDM1 | |
| MDM1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 56890 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NNEGVTNHTPVNENVELEHSTKVLSENVDNGLDRLLRKKAGLTVVPSYNALRNSEYQRQFVWKTSKETAPAFAANQVFHNKSQFVPPFKGNSVIHETEYKR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title