missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDM2/HDM2 Antibody (HDM2-323), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1312R
This item is not returnable.
View return policy
Description
MDM2/HDM2 Monoclonal specifically detects MDM2/HDM2 in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| MDM2/HDM2 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MDM2 | |
| Synthetic peptide conjugated to KLH, corresponding to amino acids 389-402 of Human MDM2. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Neuroscience, p53 Pathway, Phospho Specific, Tumor Suppressors | |
| 4193 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| HDM2-323 | |
| Western Blot, ELISA | |
| ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| MDM2 (HDM2-323) | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction