missing translation for 'onlineSavingsMsg'
Learn More

MDM2/HDM2 Antibody (MDM2/8221), Novus Biologicals™

Mouse Monoclonal Antibody

€ 249.00 - € 494.00

Specifications

Antigen MDM2/HDM2
Clone MDM2/8221
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18649794
View Documents
Novus Biologicals
NBP3-20419-100ug
100 μg
€ 494.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18670084
View Documents
Novus Biologicals
NBP3-20419-20ug
20 μg
€ 249.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MDM2/HDM2 Monoclonal antibody specifically detects MDM2/HDM2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

MDM2/HDM2
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Neuroscience, p53 Pathway, Phospho Specific, Tumor Suppressors
10mM PBS with 0.05% BSA
4193
IgG2 κ
Protein A or G purified
MDM2/8221
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase
Recombinant human MDM2/HDM2 protein fragment (around aa 1-300) (exact sequence is proprietary)
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.