missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDM2/HDM2, p Ser260 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 201.00 - € 479.00
Specifications
| Antigen | MDM2/HDM2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18665116
|
Novus Biologicals
NBP3-05385-100ul |
100 μg |
€ 479.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649386
|
Novus Biologicals
NBP3-05385-20ul |
20 μg |
€ 201.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MDM2/HDM2 Polyclonal antibody specifically detects MDM2/HDM2 in Human samples. It is validated for Western BlotSpecifications
| MDM2/HDM2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Neuroscience, p53 Pathway, Phospho Specific, Tumor Suppressors | |
| PBS with 50% glycerol, pH7.3. | |
| 4193 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase | |
| A synthetic phosphorylated peptide around S260 of human MDM2 (NP_001354919.1). SLDSE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title