missing translation for 'onlineSavingsMsg'
Learn More

MDM4 Monoclonal Antibody (OTI2B8), TrueMAB™, OriGene

Product Code. 17554542
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17554542 30 μL 30µL
17534542 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17554542 Supplier Origene Technologies Supplier No. TA505707S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MDM4 Monoclonal antibody specifically detects MDM4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MDM4
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MDM4
Gene Accession No. O15151
Gene Alias 4933417N07Rik; AA414968; AL023055; AU018793; AU021806; C85810; DKFZp781B1423; double minute 4 homolog; double minute 4 protein; double minute 4, human homolog of; p53-binding protein; HDMX; human homolog of; Mdm2-like p53-binding protein; MDM4; Mdm4 p53 binding protein homolog; MDM4 protein variant G; MDM4 protein variant Y; MDM4, p53 regulator; MDM4-related protein 1; Mdmx; MGC132766; MRP1; p53-binding protein Mdm4; Protein Mdm4; protein Mdmx; RP11-430C7.1; transformed mouse 3T3 cell double minute 4
Host Species Mouse
Immunogen Full length human recombinant protein of human MDM4 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4194
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.