Learn More
MDMX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16827924 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16827924 150 μL 150µL
16837924 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16827924 Supplier Proteintech Supplier No. 179141AP150UL
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rabbit Polyclonal Antibody
The MDM2 protein is the primary regulator of p53 protein stability. MDMX is an MDM2-related protein that inhibits MDM2-mediated degradation of p53 via distinct associations with MDM2. The gene that encodes MDMX (also designated MDM4) is a target for amplification in maligt gliomas. ARF interacts with MDMX to sequester MDMX within the nucleolus. This sequestration of MDMX by ARF results in an increase in p53 transactivation. In addition, expression of MDMX can reverse MDM2-targeted degradation of p53 while maintaining suppression of p53 transactivation. Like MDM2, MDMX also binds p73 and stabilizes the level of p73. Therefore, in striking contrast to p53, the half-life of p73 is increased by binding to MDM2.
Specifications
Antigen MDMX
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MDM4
Gene Accession No. O15151, O35618, Q5XIN1
Gene Alias Double minute 4 protein, HDMX, Mdm2 like p53 binding protein, MDM4, MDMX, MRP1, p53 binding protein Mdm4, Protein Mdm4, Protein Mdmx
Gene Symbols Mdm4
Host Species Rabbit
Immunogen MDMX Fusion Protein Ag12319
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17248, 304798, 4194
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG