missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDR1/ABCB1 Antibody (MDR1/8986R), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24231AF700
This item is not returnable.
View return policy
Description
MDR1/ABCB1 Monoclonal antibody specifically detects MDR1/ABCB1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MDR1/ABCB1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP | |
| Recombinant fragment (around aa500-700) of human MDR1/ABCB1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Membrane Trafficking and Chaperones | |
| 5243 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| MDR1/8986R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction