missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDR1/ABCB1 Antibody (MRK16) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 330.00 - € 835.00
Specifications
| Antigen | MDR1/ABCB1 |
|---|---|
| Clone | MRK16 |
| Dilution | Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, CyTOF-ready |
| Applications | Flow Cytometry, Immunoprecipitation, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18287460
|
Novus Biologicals
NBP2-52701-0.025MG |
0.025 mg |
€ 330.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670377
|
Novus Biologicals
NBP2-52701-0.1mg |
0.1 mg |
€ 835.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MDR1/ABCB1 Monoclonal specifically detects MDR1/ABCB1 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunofluorescence, CyTOF-ready.Specifications
| MDR1/ABCB1 | |
| Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP | |
| ABCB1 | |
| IgG1 | |
| Protein A or G purified |
| MRK16 | |
| Flow Cytometry, Immunoprecipitation, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Cancer, Membrane Trafficking and Chaperones | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| 5243 | |
| Drug resistant K-562 cells. | |
| Primary | |
| Recognizes a discontinuous extracellular epitope on P-glycoprotein. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title