missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDR1/ABCB1 Antibody (OTI1A7), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72607H
This item is not returnable.
View return policy
Description
MDR1/ABCB1 Monoclonal specifically detects MDR1/ABCB1 in Human, Mouse, Rat, Goat samples. It is validated for Immunohistochemistry.
Specifications
| MDR1/ABCB1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| ABCB1 | |
| Recombinant protein fragment corresponding to amino acids 995-1280 of human MDR1/ABCB1 (NP_000918) produced in SF9 cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Goat | |
| IgG2a |
| Immunohistochemistry | |
| OTI1A7 | |
| Immunohistochemistry | |
| ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP | |
| Mouse | |
| Affinity Purified | |
| Cancer, Membrane Trafficking and Chaperones | |
| 5243.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction