missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDR1/ABCB1 Antibody (S02-6B2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20044-50ul
This item is not returnable.
View return policy
Description
MDR1/ABCB1 Monoclonal antibody specifically detects MDR1/ABCB1 in Human samples. It is validated for Western Blot
Specifications
| MDR1/ABCB1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S02-6B2 | |
| Unconjugated | |
| ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP | |
| Recombinant protein of human MDR1/ABCB1 (Uniprot # P08183) | |
| 50 μg | |
| Cancer, Membrane Trafficking and Chaperones | |
| 5243 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction