missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDR3/ABCB4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 193.00 - € 483.00
Specifications
| Antigen | MDR3/ABCB4 |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18391867
|
Novus Biologicals
NBP3-03234-20UL |
20 μL |
€ 193.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18094206
|
Novus Biologicals
NBP3-03234-100UL |
100 μL |
€ 483.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MDR3/ABCB4 Polyclonal specifically detects MDR3/ABCB4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| MDR3/ABCB4 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 5244 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| ABC21, ATP-binding cassette sub-family B member 4, ATP-binding cassette, sub-family B (MDR/TAP), member 4, EC 3.6.3, EC 3.6.3.44, MDR2, MDR3MDR2/3, multidrug resistance protein 3, multiple drug resistance 3, P glycoprotein 3/multiple drug resistance 3, PFIC-3, P-glycoprotein 3, P-glycoprotein-3/multiple drug resistance-3, PGY3GBD1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human MDR3/ABCB4 (NP_061337.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title