Learn More
MDS028 Antibody (OTI3C4), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals NBP2-72614G
Code produit. 18144546
  • € 751.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
MDS028 Monoclonal specifically detects MDS028 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
MDS028
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
ITFG2
Full length recombinant protein of human MDS028 (NP_060933) produced in HEK293T cells.
0.1 mL
55846.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI3C4
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
integrin alpha FG-GAP repeat containing 2, integrin-alpha FG-GAP repeat-containing protein 2, MDS028