Learn More
ME2 Mouse Monoclonal Antibody, Proteintech
Product Code. p-200107822 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30234209 20 μL 20µL
30235148 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30234209 Supplier Proteintech Supplier No. 674571IG20UL
Mouse Monoclonal Antibody
The ME2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ME2. This antibody recognizes human, mouse, rat antigen. The ME2 antibody has been validated for the following applications: WB, IHC, ELISA analysis.
Specifications
Antigen ME2
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
Clone 6E6E2
Concentration 900 μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant Protein
Purification Method Protein A purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4200
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Antibody Molecular Weight 54 kDa, 65 kDa
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG2a