Learn More
Invitrogen™ MEA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140145
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GSGPAGYSYQ PLNQDPEQEE VELAPVGDGD VVADIQDRIQ ALGLHLPDPP LESEDEDEEG Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 92%.
MEA1 gene ontology annotations related to this gene include spermatogenesis.
Specifications
| MEA1 | |
| Polyclonal | |
| Unconjugated | |
| Mea1 | |
| HYS; male enhanced antigen 1; Male-enhanced antigen (H-Y structural gene); male-enhanced antigen 1; male-enhanced antigen-1; MEA; MEA1; Mea-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4201 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16626 | |
| Mea1 | |
| Recombinant protein corresponding to Human MEA1. Recombinant protein control fragment (Product #RP-109315). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.