Learn More
Kartell™ Measuring Scoops
Product Code. 10429222
Click to view available options
Capacity (Metric):
10 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
Pack of 6
Product Code. 10429222
Brand: Kartell™ 0105700
Only null left
Add to basket
Add to basket
Shaped conveniently for sampling
  • Flat bottom with beveled angles for easy cleaning
  • Suitable for foodstuff
  • Specifications
    Capacity (Metric) 10 mL
    Dimensions (L x W) 32 x 103 x 16 mm
    Material Polypropylene