Learn More
Mecamylamine hydrochloride, Thermo Scientific™
Product Code. 17790569 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 17790569
Brand: Thermo Scientific Acros 470010010
Request Bulk or Custom Format
Specifications
Chemical Name or Material Mecamylamine hydrochloride
CAS 826-39-1
Synonym 2-(Methylamino)isocamphane hydrochloride
Quantity 1 g
Melting Point >240°C (D)
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C11 H21 N . H Cl
Physical Form Powder
Color White to Almost-white
Purity ≥97.5%