missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MeCP2 Antibody, DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05541IR
This item is not returnable.
View return policy
Description
MeCP2 Monoclonal antibody specifically detects MeCP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| MeCP2 | |
| Monoclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| AUTSX3, DKFZp686A24160, MeCp-2 protein, mental retardation, X-linked 79, methyl CpG binding protein 2 (Rett syndrome), X-linked 16 | |
| Full length human recombinant MeCP2 expressed in and purified from E. coli. | |
| 0.1 mL | |
| Cancer, Chromatin Research, Neuroscience, Phospho Specific | |
| 4204 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction