missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MECP2 Monoclonal Antibody (4F11)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547334
This item is not returnable.
View return policy
Description
MECP2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
MECP2 belongs to a family of nuclear proteins (including MBD1, MBD2, MBD3 and MBD4) that have a methyl-CpG binding domain. MECP2 is capable of binding specifically to methylated DNA. DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. MECP2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.
Specifications
| MECP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5mM sodium azide | |
| P51608, Q00566, Q9Z2D6 | |
| Mecp2 | |
| Full length human recombinant MeCP2 expressed in and purified from E. coli. | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| 4F11 | |
| Unconjugated | |
| Mecp2 | |
| 1500041B07Rik; AUTSX3; D630021H01Rik; DKFZp686A24160; Mbd5; MECP2; MeCp-2 protein; methyl CpG binding protein 2; methyl-CpG binding protein 2; methyl-CpG-binding protein 2; MRX16; MRX79; MRXS13; MRXSL; PPMX; RS; RTS; RTT; testis tissue sperm-binding protein Li 41a; WBP10 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 17257, 29386, 4204 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction